5-Formyl-3-pyridinecarbonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 5-Formylnicotinonitrile

A159621-100MG A159621-10G A159621-1G A159621-250MG A159621-25G A159621-5G
76768-514EA 22.28 USD
76768-514 76768-516 76768-518 76768-520 76768-522 76768-524
5-Formyl-3-pyridinecarbonitrile 95%
5-Formyl-3-pyridinecarbonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₂O
MW: 132.119 g/mol
MDL Number: MFCD07367908
CAS Number: 70416-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR