5-Bromoimidazo[1,2-a]pyridine 96%

Supplier: Ambeed
Warning

A159396-1G A159396-5G A159396-10G
76768-502EA 22.28 USD
76768-502 76768-504 77598-078
5-Bromoimidazo[1,2-a]pyridine 96%
5-Bromoimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
MDL Number: MFCD08275698
CAS Number: 69214-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR