Benzothiazole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A159290-1G A159290-250MG A159290-5G
76768-494EA 29 USD
76768-494 76768-496 76768-498
Benzothiazole-2-carboxylic acid 95%
Benzothiazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₂S
MDL Number: MFCD02682025
CAS Number: 3622-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR