5-Amino-1-ethylpyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Ethyl-2H-pyrazol-3-ylamine, 1-ETHYL-PYRAZOL-5-AMINE, 1-Ethyl-1H-pyrazol-5-amine

A158859-10G A158859-5G
76768-490EA 71 USD
76768-490 76768-492
5-Amino-1-ethylpyrazole 97%
5-Amino-1-ethylpyrazole

Precaución: for Research Use Only

Formula: C₅H₉N₃
MW: 111.148 g/mol
MDL Number: MFCD00010849
CAS Number: 3528-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR