2-(5-Ethylpyridin-2-yl)ethanol 98%

Supplier: Ambeed
Warning

A158492-100G A158492-10G A158492-25G A158492-5G
76768-482EA 47 USD
76768-482 76768-484 76768-486 76768-488
2-(5-Ethylpyridin-2-yl)ethanol 98%
2-(5-Ethylpyridin-2-yl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MDL Number: MFCD00129040
CAS Number: 5223-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR