4,7-Dihydro-5H-thieno[2,3-c]pyridine-3,6-dicarboxylic acid-6-tert-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: 6-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid

A158352-1G A158352-100MG
76768-480EA 212.18 USD
76768-480 77600-252
4,7-Dihydro-5H-thieno[2,3-c]pyridine-3,6-dicarboxylic acid-6-tert-butyl ester 98%
4,7-Dihydro-5H-thieno[2,3-c]pyridine-3,6-dicarboxylic acid-6-tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄S
MW: 283.35 g/mol
MDL Number: MFCD08098464
CAS Number: 889939-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR