2-Bromo-4-fluoro-6-methylbenzonitrile 97%

Supplier: Ambeed
Warning

A158275-1G A158275-250MG
76768-472EA 22.28 USD
76768-472 76768-474
2-Bromo-4-fluoro-6-methylbenzonitrile 97%
2-Bromo-4-fluoro-6-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.0300 g/mol
MDL Number: MFCD07782071
CAS Number: 916792-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR