D-Glucoronic acid 98%

Supplier: Ambeed
Warning

Synonyms: D-(+)Glucuronic acid, D-Glucuronic acid, D-(+)-glucuronic acid, (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid

A158041-100G A158041-25G A158041-5G
76768-462EA 175 USD
76768-462 76768-464 76768-466
D-Glucoronic acid 98%
D-Glucoronic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₇
MDL Number: MFCD00166981
CAS Number: 6556-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR