4-Fluorophthalic acid 97%

Supplier: Ambeed
Warning

A157572-100G A157572-10G A157572-1G A157572-250MG A157572-25G A157572-5G
76768-416EA 206 USD
76768-416 76768-418 76768-420 76768-422 76768-424 76768-426
4-Fluorophthalic acid 97%
4-Fluorophthalic acid

Precaución: for Research Use Only

Formula: C₈H₅FO₄
MDL Number: MFCD06208213
CAS Number: 320-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR