4-Bromo-3-(trifluoromethyl)benzonitrile 97%
Supplier: Ambeed
|
Warning
|
A157462-100G
A157462-10G
A157462-1G
A157462-25G
A157462-500G
A157462-5G
76768-404EA
91
USD
76768-404
76768-406
76768-408
76768-410
76768-412
76768-414
4-Bromo-3-(trifluoromethyl)benzonitrile 97%
4-Bromo-3-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₈H₃BrF₃N |
MDL Number:
MFCD03095347 CAS Number: 1735-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...