4-Fluoro-o-toluonitrile 97%

Supplier: Ambeed
Warning

A157073-100G A157073-10G A157073-1G A157073-25G A157073-5G
76768-392EA 69 USD
76768-392 76768-394 76768-396 76768-398 76768-400
4-Fluoro-o-toluonitrile 97%
4-Fluoro-o-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆FN
MDL Number: MFCD03095106
CAS Number: 147754-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR