5-((tert-Butyldimethylsilyl)oxy)pentan-1-ol 95%
Supplier: Ambeed
|
Warning
|
A156614-1G
A156614-5G
A156614-100G
A156614-250MG
76768-384EA
44.56
USD
76768-384
77598-070
77625-444
77625-446
5-((tert-Butyldimethylsilyl)oxy)pentan-1-ol 95%
5-((tert-Butyldimethylsilyl)oxy)pentan-1-ol
Precaución: for Research Use Only
Formula:
C₁₁H₂₆O₂Si MW: 218.40 g/mol |
MDL Number:
MFCD00187970 CAS Number: 83067-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...