5-Bromo-2-methyl-2-pentene 97%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-2-methylpent-2-ene

A156418-10G A156418-1G A156418-250MG A156418-5G
76768-364EA 180.36 USD
76768-364 76768-366 76768-368 76768-370
5-Bromo-2-methyl-2-pentene 97%
5-Bromo-2-methyl-2-pentene

Precaución: for Research Use Only

Formula: C₆H₁₁Br
MW: 163.06 g/mol
MDL Number: MFCD00009887
CAS Number: 2270-59-9
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR