4-Chloro-5-methylbenzene-1,2-diamine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-4,5-diamino-2-methylbenzene, 2-Chloro-4,5-diaminotoluene

A156196-1G A156196-5G
76768-346EA 27.59 USD
76768-346 76768-348
4-Chloro-5-methylbenzene-1,2-diamine 98%
4-Chloro-5-methylbenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MW: 156.613 g/mol
MDL Number: MFCD00221471
CAS Number: 63155-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR