N(α)-Acetyl-D-tryptophan 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Trp-OH, Ac-D-Trp-OH, N-Acetyl-D-tryptophan

A155753-1G A155753-5G
76768-328EA 9.55 USD
76768-328 76768-330
N(α)-Acetyl-D-tryptophan 98%
N(α)-Acetyl-D-tryptophan

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₃
MW: 246.3 g/mol
MDL Number: MFCD00065021
CAS Number: 2280-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR