N-tert-Butyl-6-chloronicotinamide 97%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butyl)-6-chloronicotinamide

A155694-1G A155694-25G
76768-324EA 6.37 USD
76768-324 76768-326
N-tert-Butyl-6-chloronicotinamide 97%
N-tert-Butyl-6-chloronicotinamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClN₂O
MW: 212.677 g/mol
MDL Number: MFCD08460992
CAS Number: 115309-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR