4-(1-Pyrazolyl)benzonitrile 97%

Supplier: Ambeed
Warning

A154508-250MG A154508-25G A154508-5G A154508-1G
76768-280EA 5 USD
76768-280 76768-282 76768-284 77625-366
4-(1-Pyrazolyl)benzonitrile 97%
4-(1H-Pyrazol-3-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₇N₃
MDL Number: MFCD09033533
CAS Number: 25699-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR