5,6,7,8-Tetrahydro-2-methylquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 5,6,7,8-Tetrahydroquinaldine

A154305-1G A154305-250MG
76768-272EA 22.28 USD
76768-272 76768-274
5,6,7,8-Tetrahydro-2-methylquinoline 98%
5,6,7,8-Tetrahydro-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
MDL Number: MFCD00191581
CAS Number: 2617-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR