6-Bromo-2-naphthyl-β-D-galactopyranoside 95%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-naphthyl b-D-galactoside, 6-Bromo-2-naphthyl β-D-galactopyranoside

A154085-250MG A154085-1G A154085-5G
76768-268EA 18 USD
76768-268 77625-342 77877-022
6-Bromo-2-naphthyl-β-D-galactopyranoside 95%
6-Bromo-2-naphthyl-β-D-galactopyranoside

Precaución: for Research Use Only

Formula: C₁₆H₁₇BrO₆
MDL Number: MFCD00064182
CAS Number: 15572-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR