5-Methoxypyridazin-3(2H)-one 95%

Supplier: Ambeed
Warning

A154067-100MG A154067-1G A154067-250MG
76768-262EA 66.84 USD
76768-262 76768-264 76768-266
5-Methoxypyridazin-3(2H)-one 95%
5-Methoxypyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
MDL Number: MFCD05664509
CAS Number: 123696-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR