6-Fluoro-1,2,3,4-tetrahydroquinoline 97%

Supplier: Ambeed
Warning

A153759-1G A153759-250MG
76768-248EA 35.01 USD
76768-248 76768-250
6-Fluoro-1,2,3,4-tetrahydroquinoline 97%
6-Fluoro-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₉H₁₀FN
MDL Number: MFCD09040997
CAS Number: 59611-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR