2-(4-Isopropylphenoxy)acetic acid 98%

Supplier: Ambeed
Warning

A153366-5G A153366-1G
76768-232EA 29.71 USD
76768-232 77625-306
2-(4-Isopropylphenoxy)acetic acid 98%
2-(4-Isopropylphenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
MDL Number: MFCD00014364
CAS Number: 1643-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR