3-Chloro-4-(4-chlorophenoxy)aniline 97%

Supplier: Ambeed
Warning

A152958-1G A152958-250MG A152958-5G
76768-206EA 15 USD
76768-206 76768-208 76768-210
3-Chloro-4-(4-chlorophenoxy)aniline 97%
3-Chloro-4-(4-chlorophenoxy)aniline

Precaución: for Research Use Only

Formula: C₁₂H₉Cl₂NO
MDL Number: MFCD00459649
CAS Number: 24900-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR