4-(tert-Butyl)-N,N-dimethylaniline 98%

Supplier: Ambeed
Warning

Synonyms: N-[4-(tert-Butyl)phenyl]dimethylamine

A152432-1G A152432-5G A152432-25G A152432-100G
76768-198EA 21 USD
76768-198 76768-200 77592-860 77625-248
4-(tert-Butyl)-N,N-dimethylaniline 98%
4-(tert-Butyl)-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₉N
MDL Number: MFCD00008833
CAS Number: 2909-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR