Methanesulphonic anhydride 97%

Supplier: Ambeed
Danger

A152341-10G A152341-25G A152341-5G
76768-192EA 8 USD
76768-192 76768-194 76768-196
Methanesulphonic anhydride 97%
Methanesulphonic anhydride

Precaución: for Research Use Only

Formula: C₂H₆O₅S₂
MDL Number: MFCD00007556
CAS Number: 7143-01-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR