6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%

Supplier: Ambeed
Warning

A152331-10G A152331-1G A152331-250MG A152331-5G
76768-182EA 15.92 USD
76768-182 76768-184 76768-186 76768-188 76768-190
6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%
6-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁BrClN
MDL Number: MFCD08234662
CAS Number: 215798-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR