6-Isoquinolinecarboxylic acid 97%

Supplier: Ambeed
Warning

A151820-1G A151820-250MG A151820-5G
76768-168EA 38 USD
76768-168 76768-170 76768-172
6-Isoquinolinecarboxylic acid 97%
6-Isoquinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MDL Number: MFCD09910323
CAS Number: 106778-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR