6-Bromo-5-methyl[1,2,4]triazolo[1,5-a]pyridine 98%

Supplier: Ambeed
Warning

A150582-100MG A150582-250MG A150582-1G
76768-152EA 56.23 USD
76768-152 76768-154 77600-134
6-Bromo-5-methyl[1,2,4]triazolo[1,5-a]pyridine 98%
6-Bromo-5-methyl[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
MDL Number: MFCD09038165
CAS Number: 746668-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR