3,4-Dimethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A150519-100G A150519-10G A150519-1G A150519-25G A150519-5G A150519-500G
76768-142EA 114 USD
76768-142 76768-144 76768-146 76768-148 76768-150 77624-744
3,4-Dimethoxyphenylboronic acid 98%
3,4-Dimethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄
MDL Number: MFCD01074574
CAS Number: 122775-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR