Ethyl 3-(3-bromophenyl)-3-oxopropanoate 95%

Supplier: Ambeed
Warning

A149845-10G A149845-1G A149845-250MG A149845-25G A149845-5G
76768-120EA 248.26 USD
76768-120 76768-122 76768-124 76768-126 76768-128
Ethyl 3-(3-bromophenyl)-3-oxopropanoate 95%
Ethyl 3-(3-bromophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₃
MW: 271.11 g/mol
MDL Number: MFCD03424812
CAS Number: 21575-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR