5-Aminoisoquinolin-1(2H)-one 97%

Supplier: Ambeed
Warning

A149710-250MG A149710-100MG A149710-1G A149710-5G
76768-112EA 38 USD
76768-112 77877-008 77877-009 77877-010
5-Aminoisoquinolin-1(2H)-one 97%
5-Aminoisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD06198896
CAS Number: 93117-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR