5-Methyl-2-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A149397-100MG A149397-1G A149397-250MG
76768-094EA 27 USD
76768-094 76768-096 76768-098
5-Methyl-2-nitrobenzaldehyde 98%
5-Methyl-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₃
MDL Number: MFCD08703362
CAS Number: 5858-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR