5-(4-Phenoxybutoxy)psoralen 97%

Supplier: Ambeed
Warning

Synonyms: PAP-1, 4-(4-Phenoxybutoxy)-7H-furo[3,2-g]chromen-7-one

A148745-10MG A148745-1G A148745-1MG A148745-250MG A148745-25MG A148745-50MG A148745-5MG
76768-072EA 73.21 USD
76768-072 76768-074 76768-076 76768-078 76768-080 76768-082 76768-084
5-(4-Phenoxybutoxy)psoralen 97%
5-(4-Phenoxybutoxy)psoralen

Precaución: for Research Use Only

Formula: C₂₁H₁₈O₅
MW: 350.36 g/mol
MDL Number: MFCD11114059
CAS Number: 870653-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR