5-Bromo-2-phenylpyridine 97%

Supplier: Ambeed
Warning

A147877-1G A147877-250MG A147877-25G A147877-5G
76768-038EA 60.48 USD
76768-038 76768-040 76768-042 76768-044
5-Bromo-2-phenylpyridine 97%
5-Bromo-2-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₈BrN
MW: 234.10 g/mol
MDL Number: MFCD04114181
CAS Number: 27012-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR