5-Bromo-o-toluonitrile 98%

Supplier: Ambeed
Warning

A146083-100G A146083-10G A146083-25G A146083-5G
76767-998EA 143 USD
76767-998 76768-000 76768-002 76768-004 76768-006
5-Bromo-o-toluonitrile 98%
5-Bromo-o-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
MDL Number: MFCD08061920
CAS Number: 156001-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR