5-Sulfosalicylic acid dihydrate 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-5-sulphobenzoic acid dihydrate

A145193-100G A145193-500G
76767-968EA 8.49 USD
76767-968 76767-970
5-Sulfosalicylic acid dihydrate 97%
5-Sulphosalicylic acid dihydrate

Precaución: for Research Use Only

Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
MW: 254.22 g/mol
MDL Number: MFCD00149540
CAS Number: 5965-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR