2,3,3-Trimethyl-3H-indol-5-amine 95%

Supplier: Ambeed
Warning

A144966-100MG A144966-250MG A144966-5G
76767-966EA 35 USD
76767-966 77624-202 77876-988
2,3,3-Trimethyl-3H-indol-5-amine 95%
2,3,3-Trimethyl-3H-indol-5-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂
MW: 174.24 g/mol
MDL Number: MFCD00614246
CAS Number: 773-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR