2-Bromo-5-fluorobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A143953-100G
A143953-10G
A143953-1G
A143953-25G
A143953-500G
A143953-5G
76767-940EA
60
USD
76767-940
76767-942
76767-944
76767-946
76767-948
76767-950
2-Bromo-5-fluorobenzaldehyde 97%
2-Bromo-5-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄BrFO |
MDL Number:
MFCD00142872 CAS Number: 94569-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...