2,3-Dichloro-6-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A143722-100G A143722-10G A143722-25G A143722-5G
76767-926EA 177 USD
76767-926 76767-928 76767-930 76767-932
2,3-Dichloro-6-nitrobenzonitrile 97%
2,3-Dichloro-6-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂Cl₂N₂O₂
MDL Number: MFCD00728596
CAS Number: 2112-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR