5-Ethylthiazol-2-amine 97%

Supplier: Ambeed
Warning

A143645-250MG A143645-1G
76767-922EA 56.23 USD
76767-922 77592-698
5-Ethylthiazol-2-amine 97%
5-Ethylthiazol-2-amine

Precaución: for Research Use Only

Formula: C₅H₈N₂S
MW: 128.19 g/mol
MDL Number: MFCD00613421
CAS Number: 39136-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR