4-tert-Butyl-o-cresol 97%

Supplier: Ambeed
Danger

A143227-1G A143227-25G
76767-898EA 28 USD
76767-898 76767-900
4-tert-Butyl-o-cresol 97%
4-tert-Butyl-o-cresol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O
MDL Number: MFCD00020047
CAS Number: 98-27-1
UN: 2430
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR