2,5-Dibromo-3-octylthiophene 98%

Supplier: Ambeed
Warning

A143137-100G A143137-10G A143137-1G A143137-25G A143137-5G
76767-888EA 631.24 USD
76767-888 76767-890 76767-892 76767-894 76767-896
2,5-Dibromo-3-octylthiophene 98%
2,5-Dibromo-3-octylthiophene

Precaución: for Research Use Only

Formula: C₁₂H₁₈Br₂S
MW: 354.14 g/mol
MDL Number: MFCD00672013
CAS Number: 149703-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR