5-Bromo-3-nitro-o-toluic acid 97%

Supplier: Ambeed
Warning

A142369-100G A142369-10G A142369-1G A142369-250MG A142369-25G A142369-5G
76767-846EA 142 USD
76767-846 76767-848 76767-850 76767-852 76767-854 76767-856
5-Bromo-3-nitro-o-toluic acid 97%
5-Bromo-3-nitro-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
MDL Number: MFCD07357316
CAS Number: 107650-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR