N4-Benzoylcytosine 97%

Supplier: Ambeed
Warning

A142298-100G A142298-25G A142298-1000G A142298-500G
76767-844EA 15 USD
76767-844 77299-288 77624-184 77624-186
N4-Benzoylcytosine 97%
N4-Benzoylcytosine

Precaución: for Research Use Only

Formula: C₁₁H₉N₃O₂
MDL Number: MFCD00239434
CAS Number: 26661-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR