Ethyl 5-amino-1,3,4-thiadiazole-2-carboxylate 97%

Supplier: Ambeed
Warning

A142073-100G A142073-10G A142073-1G A142073-250MG A142073-25G A142073-5G
76767-814EA 930.41 USD
76767-814 76767-816 76767-818 76767-820 76767-822 76767-824
Ethyl 5-amino-1,3,4-thiadiazole-2-carboxylate 97%
Ethyl 5-amino-1,3,4-thiadiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂S
MW: 173.193 g/mol
MDL Number: MFCD03093787
CAS Number: 64837-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR