EX 527 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide, EX-527, Selisistat

A142021-1MG A142021-5MG A142021-10MG A142021-250MG A142021-50MG
76767-810EA 19.1 USD
76767-810 76767-812 77624-178 77624-180 77624-182
EX 527 98%
EX 527

Precaución: for Research Use Only

Formula: C₁₃H₁₃ClN₂O
MW: 248.71 g/mol
MDL Number: MFCD03009471
CAS Number: 49843-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR