Ac-Lys(Z)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Ac-Lys(Cbz)-OH

A141586-1G A141586-250MG A141586-5G
76767-784EA 24 USD
76767-784 76767-786 76767-788
Ac-Lys(Z)-OH 97%
Ac-Lys(Z)-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₅
MW: 322.4 g/mol
MDL Number: MFCD00136799
CAS Number: 6367-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR