3-(Trifluoromethyl)-5-isoxazolamine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-3-(trifluoromethyl)isoxazole

A140658-250MG A140658-5G A140658-1G
76767-766EA 57.29 USD
76767-766 77597-152 77624-170
3-(Trifluoromethyl)-5-isoxazolamine 95%
3-(Trifluoromethyl)-5-isoxazolamine

Precaución: for Research Use Only

Formula: C₄H₃F₃N₂O
MW: 152.08 g/mol
MDL Number: MFCD04972407
CAS Number: 108655-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR