4-sec-Butylaniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-(sec-Butyl)aniline

A140395-100G A140395-1G A140395-250MG A140395-25G A140395-5G
76767-740EA 475 USD
76767-740 76767-742 76767-744 76767-746 76767-748
4-sec-Butylaniline 98%
4-sec-Butylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₅N
MDL Number: MFCD00010223
CAS Number: 30273-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR