5-Methyl-1H-benzimidazol-2-amine 97%

Supplier: Ambeed
Danger

A139603-100MG A139603-1G A139603-250MG A139603-5G
76767-716EA 23 USD
76767-716 76767-718 76767-720 76767-722
5-Methyl-1H-benzimidazol-2-amine 97%
5-Methyl-1H-benzimidazol-2-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MDL Number: MFCD01658297
CAS Number: 6285-68-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR